Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525220

Catalog No. PIPA525220


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

UTP6 involved in nucleolar processing of pre-18S ribosomal RNA.
Specifications

Specifications

UTP6
Polyclonal
PBS with 0.09% sodium azide
Q9NYH9
C17orf40, HCA66, U3 small nucleolar RNA-associated protein 6 homolog, hepatocellular carcinoma associated antigen 66, hepatocellular carcinoma-associated antigen 66, multiple hat domains protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55813
Flow Cytometry, Western Blot
Unconjugated
UTP6
Liquid
UTP6
KLH conjugated synthetic peptide between 16-43 amino acids from the N-terminal region of human UTP6
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit