Learn More
Valpha 2 TCR Rat anti-Mouse, Brilliant Violet 605, Clone: B20.1, BD
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 563286
Catalog No. BDB563286
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The B20.1 monoclonal antibody specifically binds to most members of the V α 2 T-cell Receptor (TCR) subfamily in mice having the a, b, and c haplotypes of the Tcrb α haplotypes. Moreover, the frequencies of V α 2+ CD8+ and CD4+ T cells are influenced by H-2 haplotypes.
Specifications
Specifications
Valpha 2 TCR
B20.1
Brilliant Violet 605
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG2a, λ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
T cell receptor alpha, variable region 2
Affinity Purified
Soluble ?β TCR from mouse cytotoxic T-cell clone KB5-C20
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal