Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1C2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522131

Catalog No. PIPA522131


Add to cart

Description

Description

PA5-22131 targets V-ATPase C2 in WB applications and shows reactivity with Human samples. The PA5-22131 immunogen is a recombit fragment corresponding to a region within amino acids 3 and 237 of V-ATPase C2 (Uniprot IDNo.Q8NEY4). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

ATP6V1C2
Polyclonal
Unconjugated
ATP6V1C2
Liquid
ATP6V1C2
Recombit fragment corresponding to a region within amino acids 3 and 237 of V-ATPase C2 (Uniprot IDNo.Q8NEY4)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8NEY4
ATP6C2, VMA5, V-ATPase C2 subunit, V-ATPase subunit C 2, V-type proton ATPase subunit C 2, vacuolar proton pump subunit C 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
245973
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit