Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP6V1F Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527516

Catalog No. PIPA527516


Add to cart

Description

Description

PA5-27516 targets V-ATPase F in WB applications and shows reactivity with Human samples. The PA5-27516 immunogen is recombit fragment corresponding to a region within amino acids 1 and 119 of Human V-ATPase F. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c ft., c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.
Specifications

Specifications

ATP6V1F
Polyclonal
Unconjugated
ATP6V1F
Liquid
ATP6V1F
Recombit fragment corresponding to a region within amino acids 1 and 119 of Human V-ATPase F
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q16864
ATP6S14, VATF, Vma7, ATPase, vacuolar, 14 kD, H(+)-transporting two-sector ATPase, 14kD subunit, V-ATPase 14 kDa subunit, V-ATPase F subunit, V-ATPase subunit F, V-type proton ATPase subunit F, adenosinetriphosphatase 14k chain, vacuolar ATP synthase subunit F, vacuolar proton pump F
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9296
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit