Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-V1a Vasopressin R/AVPR1A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP144150

Catalog No. NBP144150


Add to cart

Description

Description

V1a Vasopressin R/AVPR1A Polyclonal specifically detects V1a Vasopressin R/AVPR1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

V1a Vasopressin R/AVPR1A
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry-Paraffin 1:10-1:500
Affinity Purified
AVPR1A
Synthetic 18 amino acid peptide from cytoplasmic domain of human V1a Vasopressin R/AVPR1A.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for AVPR1A. Unless other species reactivities are specified there is no testing data currently available.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.1% Sodium Azide
Antidiuretic hormone receptor 1a, arginine vasopressin receptor 1A, AVPR V1a, AVPR1, SCCL vasopressin subtype 1a receptor, V1a vasopressin receptor, V1aR, V1-vascular vasopressin receptor AVPR1A, Vascular/hepatic-type arginine vasopressin receptor, vasopressin V1a receptor
Rabbit
IgG
0.05 ml
GPCR
Primary
552
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only