Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VN1R1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA534308
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Chimpanzee, Orangutan (94%).
VN1R1 Polyclonal antibody specifically detects VN1R1 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
VN1R1 | |
Polyclonal | |
Unconjugated | |
VN1R1 | |
G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9GZP7 | |
VN1R1 | |
Synthetic 17 amino acid peptide from 2nd extracellular domain of human VN1R1. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction