Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Antibody, Agarose, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130442
Description
V5 Epitope Tag Polyclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Immunohistochemistry, Immunoprecipitation.Specifications
V5 Epitope Tag | |
Polyclonal | |
Immunohistochemistry, Immunoprecipitation 1:10-1:500 | |
V5 epitope tag | |
Rabbits were immunized with a synthetic peptide representing amino acid residues 95 to 108 of RNA polymerase alpha subunit of simion virus 5 conjugated to KLH. | |
0.1 mg | |
Epitope Tags, V5 Epitope Tags | |
Store at 4C. Do not freeze. | |
Epitope Tag |
Immunohistochemistry, Immunoprecipitation | |
Agarose | |
PBS with 0.09% Sodium Azide | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction