Learn More
UbpbioSupplier Diversity Partner ANTI-V5, MOUSE 100UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner Y1081
Antibodies – Anti-V5 (UBPBio catalog# Y1081) – Mouse monoclonal antibody (clone V5.E10, IgG1). Used for immunoblotting at 1:1,000-3,000 dilution for detection of 2 ng V5-tagged proteins. 1:500-2,000 dilution for immunostaining, 1-3 ug V5 antibody/reaction for immunoprecipitation.
Catalog No. 50-112-6923