Learn More
Vaccinia Virus A27L Mouse anti-Virus, Clone: J97Q, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518276
Catalog No. PIMA518276
Add to cart
Description
Description
This antibody is specific to the A27L protein, and is reactive with free virus and infected cells. It has tested positive with Lister and NYCBH strains of vaccinia. Purity is >90%.
Vaccinia virus is a large DNA virus with a genome of ~191 kb encoding 260 open reading frames (ORFs) that is a close relative of variola virus. Vaccinia virus morphogenesis is a complex process which occurs in the cytoplasm of infected cells and results in the formation of the intracellular mature virus (IMV) and the intracellular enveloped virus (IEV). The IMV core contains five major protein, A3L, A4L, and A10L, while 12 proteins, A12L, A13L, A14L, A14.5L, A17L, A27L, D8L, G4L, G7L, H3L, I5L and L1R, are associated with the membranes around the virus particle.
Specifications
Specifications
Vaccinia Virus A27L
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
IgG2a
100 μg
4° C
Primary
Virus
ELISA, Immunofluorescence
J97Q
Unconjugated
Liquid
Vaccinia virus native protein
Protein A
RUO
Antibody
Monoclonal