Learn More
VAChT Goat anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Manufacturer:  Invitrogen OST00046A
Catalog No. PIOST00046A
Add to cart
Description
Description
Reconstitute with 50 µL of distilled water.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Specifications
Specifications
VACHT
Polyclonal
PBS with no preservative
Q16572, O35304, Q62666
solute carrier family 18 member 3; vesicular acetylcholine transporter; SLC18A3
Goat
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20508, 60422, 6572
Immunohistochemistry, Western Blot
Unconjugated
SLC18A3
Lyophilized
SLC18A3
A synthetic peptide from human VACHT conjugated to an immunogenic carrier protein
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat