Learn More
VAChT Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136167
Catalog No. 01-672-116
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For immunohistochemistry (paraffin) applications, it is recommended to microwave samples prior to staining. For flow cytometry, incubate overnight at 4°C. Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
VACHt is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Specifications
Specifications
VAChT
Polyclonal
whole serum with 0.1% sodium azide
O35304, Q62666
VAChT, VAT
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Slc18a3
Lyophilized
SLC18A3
Synthetic peptide from the C-terminal of rat VAChT conjugated to BSA.
50 μL
4° C
Primary
20508, 60422