Learn More
VAChT Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136168
Catalog No. 01-672-117
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
VACHt is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Specifications
Specifications
VAChT
Polyclonal
whole serum with no preservative
Q16572
MGC12716, VACHT
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
SLC18A3
Lyophilized
SLC18A3
Synthetic peptide from the C-terminal of human VAChT conjugated to BSA.
50 μL
4° C
Primary
6572