Learn More
anti-VAMP-5, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130764
Catalog No. NBP130764
Add to cart
Description
Description
VAMP-5 Polyclonal antibody specifically detects VAMP-5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
VAMP-5
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
VAMP5
A synthetic peptide from the c-terminal region of human Vesicle associated membrane protein 5 (VAMP5, VAMP-5, Myobrevin) conjugated to an immunogenic carrier protein was used as the immunogen.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for VAMP5.
Human
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Sterile water with No Preservative
myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin)
Rabbit
IgG
0.5 mg
Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission
Primary
10791
Reconstitute with 0.5 ml sterilized water.