Learn More
VAMP1 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSS00038W
Catalog No. PIOSS00038W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
VAMP1
Polyclonal
whole serum with no preservative
Q62442, Q63666
VAMP-1, Vamp1, Synaptobrevin-1, Syb1
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
Vamp1
Lyophilized
VAMP1
A synthetic peptide from aa region 2-35 of mouse VAMP-1 conjugated to an immunogenic carrier protein was used as the antigen
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22317, 25624