Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OSS00047G

Catalog No. PIOSS00047G


Add to cart

Description

Description

Glycerol (1:1) may be added for additional stability. Reconstitute in 500 µL of distilled water.

Exocytosis of vesicle contents requires fusion of opposing membrane layers, and the highly conserved proteins of the SNARE family are crucially involved in this process. SNAREs localized to the vesicle membrane, or v-SNAREs, include VAMP7, and SNAREs localized to the target membrane, or t-SNARES, include the SNAPs (see SNAP25; MIM 600322) and the syntaxins (see STX1A; MIM 186590). The SNARE proteins assemble into a core complex of bundled helices and thereby contribute to membrane fusion (Sander et al., 2008 [PubMed 18253931]).
Specifications

Specifications

VAMP7
Polyclonal
Unconjugated
VAMP7
Lyophilized
VAMP7
A synthetic peptide from aa region 50-100 of human VAMP7 conjugated to an immunogenic carrier protein was used as the antigen
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
P51809, P70280, Q9JHW5
Vesicle-associated membrane protein 7, VAMP7, SYBL1, VAMP-7, Synaptobrevin-like protein 1, Tetanus-insensitive VAMP, Ti-VAMP
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20955, 6845, 85491
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit