Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vang-like Protein 2/VANGL2 Rabbit anti-Human, Mouse, Rat, Bovine, Canine, Primate, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP224704

 View more versions of this product

Catalog No. NBP224704


Add to cart

Description

Description

Vang-like Protein 2/VANGL2 Polyclonal specifically detects Vang-like Protein 2/VANGL2 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Vang-like Protein 2/VANGL2
0.5 mg/mL
Western Blot 4-6 ug/ml, Immunohistochemistry-Paraffin
Purified
VANGL2
A portion of amino acids 300-350 of human Van Gogh like-2 protein was used as the immunogen.
Protein G purified
RUO
Primary
57216
Human, Mouse, Rat, Bovine, Canine, Primate
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
KIAA1215Loop-tail protein 1 homolog, loop-tail-associated protein, LPP1, LTAPVan Gogh-like protein 2, MGC119403, MGC119404, STB1Strabismus 1, STBM, STBM1, vang (van gogh, Drosophila)-like 2, vang, van gogh-like 2 (Drosophila), vang-like 2 (van gogh, Drosophila), vang-like protein 2
Rabbit
IgG
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
100% homologous in human isoform CRA_a, CRA_b and CRA_c).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only