Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vanilloid R1/TRPV1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
$204.00 - $482.50
Specifications
Antigen | Vanilloid R1/TRPV1 |
---|---|
Concentration | 1.0 mg/ml |
Dilution | Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP171774SS
|
Novus Biologicals
NBP171774SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP171774
|
Novus Biologicals
NBP171774 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Vanilloid R1/TRPV1 | |
Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Rabbit | |
Neuroscience, Sensory Systems | |
PBS and 30% Glycerol with 0.05% Sodium Azide | |
Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 | |
TRPV1 | |
IgG | |
Affinity Purified |
1.0 mg/ml | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
Q8NER1 | |
7442 | |
A genomic peptide made to a C-terminal region of the human TRPV1 protein (within residues 725-839). [Swiss-Prot# Q8NER1] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title