Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP33 Mouse, Unlabeled, Clone: 8, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612180

Catalog No. BDB612180


Add to cart

Description

Description

In eukaryotic cells, trafficking of membrane and secretory proteins requires an elaborate system of organelles, vesicles, and cytoskeletal structures. Proteins important for protein trafficking usually interact with one or all of these cellular structures. VAP33 (VAP-A) was identified through its ability to bind the synaptic vesicle protein synaptobrevin/VAMP-1. The structure of VAP33 includes an N-terminal domain similar to the major sperm protein from Ascaris lubricoides, a central coiled-coil domain, and a C-terminal transmembrane region. VAP33 mRNA is expressed at high levels in testis, but is also found in most other tissues. In rat neurons, VAP33 localizes to the ER and microtubules, while in many cells and tissues, VAP33 co-localizes to tight junctions along with occludin. Interestingly, 83% of VAP33 fractionates with occludin and DPPIV in the plasma membrane fraction, while only 14% fractionates in the vesicular pool. In L6 skeletal myoblasts, VAP33 colocalizes with VAMP-2, and overexpression of VAP33 attenuates insulin-dependent incorporation of GLUT4 into the plasma membrane. This effect can be suppressed by overexpression of VAMP-2. Thus, VAP33 may be involved in the trafficking of plasma membrane proteins to specific sites within the cell.

Immunofluorescence, Western Blotting

Specifications

Specifications

VAP33
8
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgM
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
VAP-A
Affinity Purified
Mouse VAP33 aa. 119-226
50μg
Cell Biology
Primary
Canine, Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit