Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAPB Monoclonal Antibody (699811), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524318
Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
VAPB Monoclonal antibody specifically detects VAPB in Rat samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
VAPB | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9Z269 | |
VAPB | |
E. coli-derived recombinant rat VAP-B Ala2-Pro132 | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Frozen), Western Blot | |
699811 | |
Unconjugated | |
VAPB | |
AI225786; AI840687; AI848259; ALS 8; ALS8; ALS8VAMP-B/VAMP-C; D2Abb2e; R75548; UNQ484/PRO983; vamp; VAMP (vesicle-associated membrane protein)-associated protein B and C; Vamp 33b; VAMP associated protein B and C; VAMP B; VAMP B VAMP C; VAMP B/VAMP C; VAMP C; Vamp33b; VAMP-associated 33 kDa protein; VAMP-associated protein 33b; VAMP-associated protein B; VAMPB; VAMP-B; VAMPB/VAMPC; VAMP-B/VAMP-C; VAMPC; VAMP-C; VAP 33b; VAP B; VAP B/VAP C; VAP C; VAP33b; Vapb; VAP-B; VAPB/VAPC; VAP-B/VAP-C; VAPC; VAP-C; vesicle-associated membrane protein, associated protein B and C; vesicle-associated membrane protein-associated protein B; Vesicle-associated membrane protein-associated protein B/C | |
Mouse | |
Protein A/G | |
RUO | |
60431 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction