Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA185958

Catalog No. PA185958


Add to cart

Description

Description

Predicted to react with bovine, dog, horse, mouse, pig, rat, rhesus monkey and sheep based on sequence homology. A recommended positive control tissue is human small bowel tissue.

Specifications

Specifications

VIP
Polyclonal
Unconjugated
VIP
Liquid
Rabbit
IgG
500 μL
4°C or -20°C if preferred
Primary
7432
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.09% sodium azide
P01282
VIP
Synthetic peptide corresponding to full-length mature vasoactive intestinal peptide conjugated to KLH
Protein G
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit