Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vasopressin, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Vasopressin [Arg8].
Supplier: Abnova Corporation PAB5189
Description
Sequence: CYFQNCPRGSpecifications
Vasopressin | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Vasopressin [Arg8]. | |
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
A synthetic peptide (conjugated with KLH) corresponding to vasopressin [Arg8]. | |
RUO | |
Human | |
Liquid |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
150 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction