Learn More
AVP Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA129118
Catalog No. PIPA129118
Add to cart
Description
Description
PA1-29118 detects Vasopressin from human, and mouse samples. PA1-29118 has been successfully used in immunohistochemistry (paraffin tissue), and Western blot applications. The PA1-29118 immunogen is synthetic peptide corresponding to the C-terminus of human vasopressin. Positive Control: Kidney tubule
Avpr2 is a precursor protein consisting of arginine vasopressin and two associated proteins, neurophysin 2 and a glycopeptide, copeptin. Arginine vasopressin is a posterior pituitary hormone which is synthesized in the supraoptic nucleus and paraventricular nucleus of the hypothalamus. Along with its carrier protein, neurophysin 2, it is packaged into neurosecretory vesicles and transported axonally to the nerve endings in the neurohypophysis where it is either stored or secreted into the bloodstream. The precursor is thought to be activated while it is being transported along the axon to the posterior pituitary. Arginine vasopressin acts as a growth factor by enhancing pH regulation through acid-base transport systems. Avpr2 has a direct antidiuretic action on the kidney, and also causes vasoconstriction of the peripheral vessels. Avpr2 can contract smooth muscle during parturition and lactation. Avpr2 is also involved in cognition, tolerance, adaptation and complex sexual and maternal behaviour, as well as in the regulation of water excretion and cardiovascular functions. The activity of Avpr2 is mediated by G proteins which activate adenylate cyclase.
Specifications
Specifications
AVP
Polyclonal
Unconjugated
AVP
Liquid
Rabbit
IgG
500 μL
4° C, do not freeze
Primary
11998, 551
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
10mM PBS with 0.2% BSA and 0.1% sodium azide; pH 7.4
P01185, P35455
AVP
Synthetic peptide corresponding to the C-terminus of human vasopressin recceptor V2
Protein A
RUO
Antibody
Polyclonal
Human, Mouse