Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vav Mouse anti-Human, Mouse, MilliporeSigma™

Mouse Monoclonal Antibody

Manufacturer:  MilliporeSigma 05219

Catalog No. 05-219-MI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Anti-Vav Antibody is an antibody against Vav for use in western blotting.
Specifications

Specifications

Vav
Unconjugated
NM_005428.2
VAV1, VAV
GST-fusion to full length human Vav
200μL
Primary
Human, Murine
Western Blot
ascites fluid containing 0.05% sodium azide
Ascites
Murine
IgG
−20°C for 24 months.
Monoclonal
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only