Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAV2, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA131284

Catalog No. PIPA131284

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

VAV2 is the second member of the VAV guanine nucleotide exchange factor family of oncogenes. Unlike VAV1, which is expressed exclusively in hematopoietic cells, VAV2 transcripts were found in most tissues. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

PA1-31284 detects VAV2 from human samples.PA1-31284 has been successfully used in immunoprecipitation, Western blot applications.The PA1-31284 immunogen is: His tagged fusion protein, corresponding to amino acids 578-878 of Human VAV2.
Specifications

Specifications

VAV2
Polyclonal
PBS with 0.08% sodium azide
P52735
VAV2
His tagged fusion protein, corresponding to amino acids 578-878 of Human VAV2.
IgG fraction
RUO
Antibody
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
VAV2
Purified
Sheep
IgG
250μg
Store at -20°C
Primary
7410
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit