Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vb 13 TCR Mouse anti-Mouse, APC, Clone: MR12-3, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 561542

Catalog No. BDB561542


Add to cart

Description

Description

The MR12-3 monoclonal antibody specifically binds to the V β 13 T-cell Receptor (TCR) of mice having the b haplotype (e.g., A, AKR, BALB/c, CBA, C3H/He, C57BL, C58, DBA/1, DBA/2) of the Tcrb gene complex. The Tcrb-V13 c (e.g., RIII) haplotype.

Specifications

Specifications

Vb 13 TCR
MR12-3
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
TCR V beta 13; TCR Vβ 13; TCR Vβ13; Vβ 13 T-cell Receptor
Affinity Purified
Not reported
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit