Learn More
anti-VCAM-1/CD106, Alexa Fluor 488, Clone: IE10, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB5649G
Catalog No. FAB5649G
Add to cart
This item is not returnable. View return policy
Description
Description
VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
VCAM-1/CD106
IE10
Flow Cytometry 5 uL/10^6 cells
Purified
VCAM1
Activated HUVEC human umbilical vein endothelial cells
Protein A or G purified from ascites
RUO
Primary
7412
Human
Flow Cytometry
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
The antibody was screened using COS cells transfected with cDNAs for E-Selectin, VCAM-1, ICAM-1 and PECAM-1 and was shown to be specific for human VCAM-1/CD106.