Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF 165b Mouse anti-Human, Clone: 56-1, R&D Systems™

Mouse Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen VEGF
Immunogen KLH-conjugated human VEGF 165b synthetic peptide TCRSLTRKD Accession # AAL27435
Clone 56-1
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB3045SP
View Documents
R&D Systems
MAB3045SP
25 ug
Each for $114.50
Add to cart
 
MAB3045100
View Documents
R&D Systems
MAB3045100
100 ug
Each for $377.00
Add to cart
 
MAB3045500
View Documents
R&D Systems
MAB3045500
500 ug
Each for $839.00
Add to cart
 
Description

Description

VEGF 165b Monoclonal antibody MAB3045 Specifically detects VEGF 165b in Human samples. It is validated for Immunofluorescence, Immunohistochemistry, Western Blotting
Specifications

Specifications

VEGF
56-1
Unconjugated
AAL27435
IgG1
7422
RUO
KLH-conjugated human VEGF 165b synthetic peptide TCRSLTRKD Accession # AAL27435
Western Blot, Immunohistochemistry, Immunofluorescence
Purified
Monoclonal
Primary
Detects human VEGF 165b in direct ELISAs and Western blots. Recognizes an epitope within a 9 amino acid sequence at the C-terminus of human VEGF 165b . In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) VEGF 206 rhVEGF-B 167 rhVEGF-B 186 rhVEGF-C, or rhVEGF-D is observed.
Protein A or G purified from hybridoma culture supernatant
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit