Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGFC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen 345500

Encompass

Catalog No. 34-550-0


Add to cart

Description

Description

The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.
Specifications

Specifications

VEGFC
Polyclonal
Unconjugated
VEGFC
Liquid
VEGFC
Synthetic peptide derived from the internal region of the human VEGF-C protein
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.25 mg/mL
PBS with 0.1% sodium azide; pH 7.4
P49767
Flt4-L, VRP, FLT4 ligand DHM, VEGF-C, vascular endothelial growth factor-related protein
Rabbit
IgG
100 μg
-20°C
Primary
7424
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit