Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF R1/Flt-1 Rat anti-Mouse, Clone: 141515, R&D Systems™

Rat Monoclonal Antibody

$114.50 - $839.00

Specifications

Antigen VEGFR1/Flt-1
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1, Ser27-Glu759, Accession # P35969
Clone 141515
Applications Western Blot, Immunohistochemistry
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB471SP
View Documents
R&D Systems
MAB471SP
25 ug
Each for $114.50
Add to cart
 
MAB471100
View Documents
R&D Systems
MAB471100
100 ug
Each for $377.00
Add to cart
 
MAB471500
View Documents
R&D Systems
MAB471500
500 ug
Each for $839.00
Add to cart
 
Description

Description

VEGF R1/Flt-1 Monoclonal antibody MAB471 Specifically detects VEGF R1/Flt-1 in Mouse samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

VEGFR1/Flt-1
141515
Unconjugated
P35969
Monoclonal
Primary
Detects mouse VEGFR1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human VEGFR1, recombinant mouse (rm) VEGFR2, or rmVEGFR3 is observed.
Protein A or G purified from hybridoma culture supernatant
Mouse myeloma cell line NS0-derived recombinant mouse VEGFR1, Ser27-Glu759, Accession # P35969
Western Blot, Immunohistochemistry
Purified
Rat
IgG2a
2321
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit