Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human VEGFR3/Flt-4 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
$172.84 - $1184.00
Specifications
Antigen | VEGFR3/Flt-4 |
---|---|
Clone | 54716 |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Classification | Monoclonal |
Description
VEGFR3/Flt-4 Monoclonal specifically detects VEGFR3/Flt-4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
VEGFR3/Flt-4 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P35916 | |
2324 | |
Mouse myeloma cell line NS0-derived recombinant human VEGFR3, Tyr25-Ile776, Accession # P35916 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human VEGFR3 in ELISAs. In ELISAs, shows 25% cross-reactivity with recombinant mouse (rm) VEGFR3 and no cross-reactivity with rhVEGFR1 or rhVEGFR2. |
54716 | |
ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.7.10, EC 2.7.10.1, Flt-4, FLT4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGF R3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4 | |
FLT4 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title