Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VEGF Receptor 2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Catalog No. PIPA132431
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA132431 100 μL
1 options

Catalog No. PIPA132431

Supplier: Thermo Scientific PA132431

Rabbit Polyclonal Antibody

PA1-32431 detects VEGFR2 from mouse samples. PA1-32431 has been successfully used in immunoprecipitation, Western blot applications. The PA1-32431 immunogen is: This antibody was made to residues corresponding to the kinase insert or C-terminus of mouse VEGFR-2 of (residues 1158-1365).

TRUSTED_SUSTAINABILITY

Specifications

Antigen VEGF Receptor 2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene KDR
Gene Accession No. P35968, P35918
Gene Alias 6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR
Gene Symbols KDR
Host Species Rabbit
Immunogen This antibody was made to residues corresponding to the kinase insert or C-terminus of murine VEGFR-2 of (residues 1158-1365).
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16542, 3791
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.