Learn More
VEGF Receptor 2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137708
Description
Flk-1 / KDR / VEGFR2, a cell membrane receptor kinase, is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis. It contains seven immunoglobulin like sequences in the extracellular domain and exhibits sequence similarity to Flt-1 and Flt-4.
This antibody is predicted to react with mouse based on sequence homology.Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta or angiosarcoma.
Specifications
VEGF Receptor 2 | |
Polyclonal | |
KDR | |
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2, CD309, KDR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3791 | |
Store at 4°C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
P35968 | |
KDR | |
Synthetic peptide derived from C-terminus of mouse Flk-1 | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.