Learn More
VEGF Receptor 3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137711
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is placenta or breast carcinoma.
Three cell membrane receptor tyrosine kinases, Flt (also designated VEGF-R1), Flk-1 (also designated VEGF-R2) and Flt-4 (also designated VEGF-R3), putatively involved in the frowth of endothelial cells, are characterized by the presence of seven immuno-globulin-like sequences in their extracellular domain. On the basis of structural similarity to Flt and Flk-1, it has been speculated that Flt-4 might represent a third receptor for either CEGF or VEGF-related ligand.
Specifications
VEGF Receptor 3 | |
Polyclonal | |
FLT4 | |
FLT41, PCL, VEGFR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2324 | |
4° C | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P35916 | |
FLT4 | |
Synthetic peptide derived from C-terminus of human Flt-4 protein | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.