Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VENTX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521006
Description
A suggested positive control is mouse brain tissue lysate. PA5-21006 can be used with blocking peptide PEP-1120.
VENTX Polyclonal antibody specifically detects VENTX in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
VENTX | |
Polyclonal | |
Unconjugated | |
Ventx | |
hemopoietic progenitor homeobox protein VENTX2; homeobox protein VENTX; HPX42B; NA88A; VENT homeobox; VENT homeobox homolog; VENT-like homeobox 2; VENT-like homeobox protein 2; VENTX; VENTX2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
110333234, 27287 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O95231 | |
Ventx | |
A 16 amino acid peptide near the center of human VENTX. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction