Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Shiga toxin 2B, clone: F410C, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA17098

Catalog No. MA17098

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MA1-7098 detects the Verotoxin. This antibody is specific for Verotoxin II - B subunit (SLT-2b).MA1-7098 has been successfully used in ELISA procedures.The MA1-7098 immunogen is purified native toxoid.
Specifications

Specifications

Verotoxin
Monoclonal
0.1mg/mL
PBS with 0.1% sodium azide; pH 7.2
Shiga toxin, STX-IIb, STX-2b, Shigatoxin Type 2, Shigatoxin Type II, STX2
Purified native toxoid.
Protein A
RUO
Antibody
Monoclonal
ELISA
F410C
Unconjugated
Purified
Murine
IgG
1mL
Store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit