Learn More
MilliporeSigma™ anti-Vesicular Acetylcholine Transporter Ab-2; Polyclonal; ™ (Chemicon™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB1588/DEL
Catalog No. 50-173-188
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Vesicular Acetylcholine Transporter (VAChT). By Western blot the antibody reacted with a single band in rat brain homogenates. This band corresponded to the size of the predicted rVAChT. There was no staining of blots incubated with the antibody that had been preabsorbed with the peptide used for antibody production. By immunohistochemistry the staining was strong in the inner retina where cholinergic processes are located and weak in cholinergic cell bodies. There was no background staining noted. In the CNS staining was strong in fibers and stains cholinergic cell bodies well.
Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Vesicular Acetylcholine Transporter Ab-2
Affinity Purified
Synthetic peptide corresponding to the C-terminus of the predicted rat VAChT protein.
RUO
Primary
Rat
Unconjugated
Guinea Pig
20μg
Neuroscience
Polyclonal