Learn More
VGAT Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136114
Catalog No. 01-674-520
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A recommended positive control for IHC (frozen sections) is rat cerebellum. Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
The Vesicular GABA Amino Acid Transporter (VGAT) is responsible for transport of the inhibitory neurotransmitter into synaptic vesicles (McIntire et al., 1997). The VGAT protein (also known as the Vesicular Inhibitory Amino Aid Transporter or VIAAT) is expressed in synaptic vesicles of both glycine and GABAergic synapses throughout the CNS (Chaudhry et al., 1998). Expression of the VGAT protein changes during development and also in response to patterns of neuronal activity (De et al., 2005).
Specifications
Specifications
VGAT
Polyclonal
whole serum with 0.1% sodium azide
O35458
Vgat, Viaat
Rabbit
IgG
RUO
Antibody
Polyclonal
Rat
Immunofluorescence, Immunohistochemistry (Frozen)
Unconjugated
Slc32a1
Lyophilized
SLC32A1
Synthetic peptide from the C-terminal of VGAT conjugated to BSA.
50 μL
4° C
Primary
83612