Learn More
VG5Q Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA140291
Catalog No. 01-676-410
Add to cart
Description
Description
PA1-40291 detects VG5Q from mouse samples. PA1-40291 has been successfully used in ELISA, immunohistochemistry (frozen), immunoprecipitation, immunohistochemistry (paraffin), and Western blot applications. The PA1-40291 imunogen is mouse VG5Q.
The VG5Q protein is a potent angiogenic factor that contains a forkhead-associated domain and a G-patch domain.
Specifications
Specifications
VG5Q
Polyclonal
Unconjugated
Aggf1
Liquid
AGGF1
Synthetic murine VG5Q peptide
Purified
RUO
Antibody
Polyclonal
Mouse
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.1 mg/mL
PBS with 0.1% BSA and 0.02% sodium azide
Q7TN31
2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q
Rabbit
IgG
100 μg
4° C
Primary
66549