Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vγ 2 T-Cell Receptor Hamster anti-Mouse, PE, Clone: UC3-10A6, BD

Hamster Monoclonal Antibody

Manufacturer:  BD Biosciences 565167

Catalog No. BDB565167


Add to cart

Description

Description

The UC3-10A6 monoclonal antibody specifically recognizes T-cell Receptor (TCR) V gamma 2 ( Vγ2 ). Vγ2 + TCR γδ cells make up significant proportions of TCR γδ cells in the late fetal and adult thymus and adult peripheral lymphoid tissues and lung. The frequency of splenic Vγ2 + TCR γδ cells differs among inbred mouse strains; in C57BL/6 mice, the frequency increases dramatically during the four weeks after birth. Immobilized UC3-10A6 antibody can activate Vγ2 + TCRγδ cells. Please note that the Vγ2 designation correlates with the nomenclature of Garman, Doherty, and Raulet; the Vγ4 designation of Heiligand Tonegawa is equilvalent.

Specifications

Specifications

Vγ 2 T-Cell Receptor
UC3-10A6
PE
Aqueous buffered solution containing ≤0.09% sodium azide
Hamster
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
V gamma 2 TCR; Vγ2 TCR
Affinity purified
Not reported
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit