Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524311

Catalog No. PIPA524311

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May act as a specific coactivator for the mammalian TEFs. May play a role in the development of skeletal muscles.
Specifications

Specifications

VGLL2
Polyclonal
PBS with 0.09% sodium azide
Q8N8G2, Q8BGW8
VGL2, VITO1, transcription cofactor vestigial-like 2, transcription cofactor vestigial-like protein 2, vestigial-like 2, vgl-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
215031, 245806
Western Blot
Unconjugated
VGLL2
Liquid
VGLL2
KLH conjugated synthetic peptide between 49-77 amino acids from the N-terminal region of human VGLL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit