Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLUT2, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525653

Catalog No. PIPA525653

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

MOUSE VGLU2 mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neural cells. This protein may also mediate the transport of inorganic phosphate.

This antibody is predicted to react with bovine and rat based on sequence homology.
Specifications

Specifications

VGLU2
Polyclonal
PBS with 0.09% sodium azide
Q9P2U8, Q8BLE7
2900073D12Rik, DNPI, VGLUT2, differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, solute carrier family 17 (vesicular glutamate transporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2
Rabbit
IgG
400μL
Store at 2°-8°C for up to 6 months. For long term storage store at -20°C
Primary
57084
Immunohistochemistry, Western Blot
Unconjugated
SLC17A6
Purified
SLC17A6
KLH conjugated synthetic peptide between 507-535 amino acids from the C-terminal region of mouse VGLU2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit