Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Full-Finger Anti-Vibration Air Glove™ with Silicone Dots

Patented Air Glove™ technology protects hands from impact and vibration. Synthetic suede leather with silicone dots on the palm & fingers provide comfort and enhanced grip abilities.

$38.82

Specifications

Coating Material Silicone
Color Black
For Use With (Application) Construction and Maintenance
Glove Cuff Style Hook and Loop
Back Material Mesh
View More Specs

Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-400-044
View Documents
Impacto™
BG40820
Small
Pair for $38.82
Add to cart
 
19-400-045
View Documents
Impacto™
BG40830
Medium
Pair for $38.82
Add to cart
 
19-400-046
View Documents
Impacto™
BG40840
Large
Pair for $38.82
Add to cart
 
19-400-047
View Documents
Impacto™
BG40850
X-Large
Pair for $38.82
Add to cart
 
19-400-048
View Documents
Impacto™
BG40860
2X-Large
Pair for $38.82
Add to cart
 
Description

Description

  • Synthetic ultra suede leather provides extended wear and flexibility
  • Silicone dots on the palm and fingertips enhance grip capabilities
  • Mesh back allows ventilation and comfort
  • Durable Bubble Glove™ bladder in the palm, fingers, and thumb protects from vibration
  • Full finger style offers maximum protection and tactile feedback
  • Neoprene cuff with hook and loop closure
  • Vibration protection when using hand-held or hand guided power tools and machines
Specifications

Specifications

Silicone
Construction and Maintenance
Mesh
Anti-Vibration Gloves
Black
Hook and Loop
Synthetic Leather
BG408
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit