Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HDLBP Rabbit anti-Canine, Equine, Human, Mouse, Non-human primate, Porcine, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523310

Catalog No. PIPA523310


Add to cart

Description

Description

Vigilin, also known as high density lipoprotein binding protein (HDLBP), is a 1268 amino acid containing cytoplasmic protein in close association with the endoplasmic reticulum with fourteen K homology (KH).  A transport protein, widely expressed in most of tissues, appears to play a role in cell sterol metabolism. It specifically binds to HDL (high density lipoprotein) molecules and may function to protect cells from over-accumulation of cholesterol by removal of excess cellular cholesterol.
Specifications

Specifications

HDLBP
Polyclonal
Unconjugated
HDLBP
Liquid
HDLBP
Synthetic peptide (526 KGERIREIRDKFPEV 540∽^925 QENGDEAGEGREAKDC 940) of Vigilin/HDLBP  protein.
Protein G
RUO
Antibody
Polyclonal
Canine, Equine, Human, Mouse, Non-human primate, Porcine, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q00341, Q8VDJ3, Q9Z1A6
HBP, PRO2900, VGL, HDL-binding protein, high density lipoprotein-binding protein, vigilin
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100057882, 100233187, 110611, 3069, 477431, 64474
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit