Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VILIP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP214988
Description
VILIP3 Polyclonal specifically detects VILIP3 in Human, Mouse samples. It is validated for Western Blot.Specifications
VILIP3 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
BDR1Visinin-like protein 3, Calcium-binding protein BDR-1, hippocalcin-like 1, HLP2hippocalcin-like protein 1, VILIP-3 | |
Rabbit | |
22 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Predicted cross-reactivity based on sequence identity: Xenopus laevis (96%) and Xenopus tropicalis (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.23mg/mL | |
Western Blot 1:500-1:3000 | |
P37235 | |
HPCAL1 | |
Recombinant protein encompassing a sequence within the center region of human HPCAL1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
3241 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction