Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Vimentin, Polyclonal, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB45939100UG

Catalog No. NC0122379


Add to cart

Description

Description

Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Cow, Human. Predicted to work with Rat, Sheep, Chicken, Pig, Chimpanzee
Immunogen: Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Human Vimentin.

Applications:

Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting

Specifications

Specifications

Vimentin
Unconjugated
Rabbit
IgG
Cancer and Tumor Biology, Developmental Biology, Neuroscience, Signal Transduction, Stem Cell Biology
Polyclonal
0.6-1mg/mL
Affinity Purified
Synthetic peptide conjugated to KLH derived from within residues 400 to the C-terminus of Human Vimentin.
100μg
Primary
Bovine, Chicken, Chimpanzee, Human, Murine, Porcine, Rat, Sheep
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit