Learn More
VIP Guinea Pig anti-Guinea Pig, Human, Porcine, Rat, Polyclonal, Invitrogen™
Guinea Pig Polyclonal Antibody
Manufacturer:  Invitrogen PA136030
Catalog No. 01-672-220
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 μL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
The protein encoded by this gene belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
Specifications
Specifications
VIP
Polyclonal
whole serum with no preservative
P01282, P01284, P01283
MGC13587, PHM27
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Guinea Pig, Human, Porcine, Rat
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
VIP
Lyophilized
VIP
Synthetic human VIP.
50 μL
4° C
Primary
100500718, 100735454, 117064, 7432