Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viperin Mouse, PE, Clone: MaP.VIP, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565196

Catalog No. BDB565196


Add to cart

Description

Description

The MaP.VIP monoclonal antibody specifically binds to Virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible (Viperin), which is also known as Radical S-adenosyl methionine domain-containing protein 2 (Rsad2). Viperin is a 43 kDa protein that is localized to the endoplasmic reticulum and lipid droplets. It functions in protective responses to a number of different DNA and RNA viruses. Viperin expression is induced in cells by different agents such as, Type I interferons, Interferon- γ(IFN- γ), DNA and RNA viral proteins, Polyinosinic:polycytidylic acid, and lipopolysaccharide. Various cell types can be induced to express Viperin including T and B lymphocytes, macrophages, granulocytes, dendritic cells, fibroblasts, endothelial cells, and epithelial cells.

Specifications

Specifications

Viperin
MaP.VIP
PE
Aqueous buffered solution containing ≤0.09% sodium azide
Mouse
IgG2a, κ
RUO
Primary
Human, Murine, Rat
Flow Cytometry
0.2mg/mL
Rsad2; Vig1; cig5
Affinity purified
Mouse Viperin Recombinant Protein
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit