Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIPR1, Polyclonal, Invitrogen™

Catalog No. PIPA132574
Change view
Click to view available options
Quantity:
25 μg
Catalog No. Quantity
PIPA132574 25 μg
1 options

Catalog No. PIPA132574

Supplier: Thermo Scientific PA132574

Rabbit Polyclonal Antibody

This gene encodes a receptor for vasoactive intestinal peptide, a small neuropeptide. Vasoactive intestinal peptide is involved in smooth muscle relaxation, exocrine and endocrine secretion, and water and ion flux in lung and intestinal epithelia. Its actions are effected through integral membrane receptors associated with a guanine nucleotide binding protein which activates adenylate cyclase.

PA1-32574 detects VIPR1 from human samples.PA1-32574 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-32574 immunogen is: Synthetic peptide, KLH conjugated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIPR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene VIPR1
Gene Accession No. P32241
Gene Alias FLJ41949, HVR1, II, PACAP-R-2, RDC1, V1RG, VAPC1, VIPR, VIRG, VPAC1, VPCAP1R
Gene Symbols VIPR1
Host Species Rabbit
Immunogen Synthetic peptide, KLH conjugated.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.