Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VKORC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525753

Catalog No. PIPA525753


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Specifications

Specifications

VKORC1
Polyclonal
PBS with 0.09% sodium azide
Q9BQB6
EDTP308, IMAGE3455200, MST134, MST576, VKCFD2, VKOR, phylloquinone epoxide reductase, vitamin K dependent clotting factors deficiency 2, vitamin K epoxide reductase complex subunit 1, vitamin K1 2,3-epoxide reductase subunit 1, vitamin K1 epoxide reductase (warfarin-sensitive)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79001
Immunohistochemistry, Western Blot
Unconjugated
VKORC1
Liquid
VKORC1
KLH conjugated synthetic peptide between 15-41 amino acids from the N-terminal region of human VKORC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit