Learn More
VMAT2 Guinea Pig anti-Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™
Guinea Pig Polyclonal Antibody
Manufacturer:  Invitrogen PA136031
Catalog No. 01-672-221
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
The vesicular monoamine transporter acts to accumulate cytosolic monoamines into synaptic vesicles, using the proton gradient maintained across the synaptic vesicular membrane. Its proper function is essential to the correct activity of the monoaminergic systems that have been implicated in several human neuropsychiatric disorders. The transporter is a site of action of important drugs, including reserpine and tetrabenazine (Peter et al., 1993 [PubMed 7905859]).
Specifications
Specifications
VMAT2
Polyclonal
whole serum with no preservative
Q05940, Q8BRU6, Q01827
MGC120477, MGC120478, MGC26538, SVAT, SVMT, VAT2, VMAT2
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Porcine, Rat
Immunohistochemistry (Frozen)
Unconjugated
SLC18A2
Lyophilized
SLC18A2
Synthetic peptide from the C-terminal of rat VMAT2 conjugated to BSA.
50 μL
4° C
Primary
100154950, 214084, 25549, 6571